Stearinsäure ≥98%
Glasflasche für Feststoffe, 5 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 57-11-4 Molecular Formula: CH3(CH2)16CO2H Formula Weight: 284.48 Flash Point: 196°(384°F) Physical Form: Flakes or powder Density: 0.941 Appearance: White Odour: Odourless Melting Point: 69-72° Boiling Point: 232°/15mm Beilstein Reference: 608585 Merck Reference: 14,8804 MDL No.: MFCD00002752
A saturated fatty acid
Synonyme: Octadecanoic acid, Octadecylic acid
- CAS Nummer:57-11-4
- Formel:C₁₈H₃₆O₂
- MDL-Nummer:MFCD00002752
- Molekulargewicht:284.48 g/mol
- Merck-Index:14,8804
- Siedepunkt:232 °C (15 mmHg)
- Schmelzender Platingehalt:69…72 °C
- Dichte:0,941
- Blitz Pt:196 °C (384 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:200-313-4
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFAA12244
Sicherheitsdatenblatt (SDS)
Organic Building Blocks and Intermediates
Anwendungshinweise
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



