Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Methyldimethoxyacetat ≥97% | Avantor

Methyldimethoxyacetat ≥97%

Glasflasche für Feststoffe, 50 g
Catalog # A12171.18
Methyldimethoxyacetat ≥97%
Catalog # A12171.18
CAS Number:  89-91-8
Methyldimethoxyacetat ≥97%
Methyldimethoxyacetat ≥97%

Glasflasche für Feststoffe, 50 g

CAS Number:  89-91-8
Price
454.00 SFr.
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 97+% CAS No.: 89-91-8 Molecular Formula: (CH3O)2CHCO2CH3 Formula Weight: 134.13 Flash Point: 63°(145°F) Refractive Index: 1.4050 Boiling Point: 160-163° Beilstein Reference: 1757582 MDL No.: MFCD00008484

Synonyms:  Methyl 2,2-dimethoxyacetate
  • CAS Number:89-91-8
  • Formula:C₅H₁₀O₄
  • MDL Number:MFCD00008484
  • Molecular Weight:134.13 g/mol
  • Boiling Pt:160…163 °C
  • Density:1,098
  • Flash Pt:63 °C (145 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:201-950-0
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.