(±)-Isonipecotinsäure ≥98%
Glass bottle for solids, 25 g
Anbieter: Thermo Fisher Scientific
(±)-Isonipecotinsäure ≥98%
Katalognr. A11795.14
Anbieter: Thermo Fisher Scientific
CAS Nummer: 498-94-2
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 498-94-2 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Odour: Odourless Melting Point: 350° Beilstein Reference: 112553 Merck Reference: 14,5189 MDL No.: MFCD00006004
Synonyme: Piperidine-4-carboxylic acid, (±)-4-Piperidinecarboxylic acid, 4-Piperidinecarboxylic acid, Isonipecotic acid
- CAS Nummer:498-94-2
- Formel:C₆H₁₁NO₂
- MDL-Nummer:MFCD00006004
- Molekulargewicht:129.16 g/mol
- Merck-Index:14,5189
- Siedepunkt:264 °C (1013 hPa)
- Schmelzender Platingehalt:>350 °C
- Lagertemperatur:Raumtemperatur
- EINECS:207-872-3
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFAA11795
Sicherheitsdatenblatt (SDS)
Organic Building Blocks and Intermediates
Anwendungshinweise
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



