Ethylenglycol ≥99%
Glasflasche für Feststoffe, 500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 107-21-1 Molecular Formula: C2H6O2 Formula Weight: 62.07 Flash Point: 119°(246°F) Refractive Index: 1.4310 Odour: Sweet Melting Point: -13° Boiling Point: 196-198° Beilstein Reference: 505945 Merck Reference: 14,3798 MDL No.: MFCD00002885
Synonyme: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
- CAS Nummer:107-21-1
- Formel:HOCH₂CH₂OH
- MDL-Nummer:MFCD00002885
- Molekulargewicht:62.07 g/mol
- Merck-Index:14,3798
- Siedepunkt:196…198 °C
- Schmelzender Platingehalt:–13 °C
- Dichte:1,113
- Blitz Pt:119 °C (246 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:203-473-3
- ERREICHEN:01-2119456816-28
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:500 g
- Cat. No.:A11591.36
- Umweltzertifizierung:Chem21 Solvent Selection Guide recommended by default and after discussion
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



