Undecansäure ≥98%
Glasflasche für Feststoffe, 250 g
Purity: 98% CAS No.: 112-37-8 Molecular Formula: CH3(CH2)9CO2H Formula Weight: 186.30 Flash Point: 110°(230°F) Melting Point: 26-29° Boiling Point: 283-285° Beilstein Reference: 1759287 MDL No.: MFCD00002730
: Hendecanoic acid, Undecylic acid
- :112-37-8
- :C₁₁H₂₂O₂
- :MFCD00002730
- :186.29 g/mol
- :283…285 °C
- :26…29 °C
- :0,89 g/cm³ (20 °C)
- :>110 °C (230 °F)
- :Raumtemperatur
- :203-964-2
Documents
- Packung:Glasflasche für Feststoffe
- VE:250 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.