Trichloressigsäure ≥99%
Glasflasche für Feststoffe, 500 g
Purity: 99% CAS No.: 76-03-9 Molecular Formula: Cl3CCO2H Formula Weight: 163.39 Odour: Acrid pH: 1 Melting Point: 54-58° Boiling Point: 196° Beilstein Reference: 970119 Merck Reference: 14,9627 UN No.: 1839 MDL No.: MFCD00004177
: TCA
- :76-03-9
- :Cl₃CCOOH
- :MFCD00004177
- :163.39 g/mol
- :14,9627
- :196 °C
- :54…58 °C
- :1,629
- :113 °C (geschlossener Tiegel)
- :Raumtemperatur
- :200-927-2
- :8,II
Documents
- Packung:Glasflasche für Feststoffe
- VE:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.