Trichloressigsäure ≥99%
Glasflasche für Feststoffe, 5.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 76-03-9 Molecular Formula: Cl3CCO2H Formula Weight: 163.39 Odour: Acrid pH: 1 Melting Point: 54-58° Boiling Point: 196° Beilstein Reference: 970119 Merck Reference: 14,9627 UN No.: 1839 MDL No.: MFCD00004177
Synonyms: TCA
- CAS Number:76-03-9
- Formula:Cl₃CCOOH
- MDL Number:MFCD00004177
- Molecular Weight:163.39 g/mol
- Merck Index:14,9627
- Boiling Pt:196 °C
- Melting Pt:54…58 °C
- Density:1,629
- Flash Pt:113 °C (geschlossener Tiegel)
- Storage Temperature:Raumtemperatur
- EINECS:200-927-2
- ADR:8,II
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:5.000 g



