Trichloressigsäure ≥99%
Glasflasche für Feststoffe, 1.000 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 76-03-9 Molecular Formula: Cl3CCO2H Formula Weight: 163.39 Odour: Acrid pH: 1 Melting Point: 54-58° Boiling Point: 196° Beilstein Reference: 970119 Merck Reference: 14,9627 UN No.: 1839 MDL No.: MFCD00004177
Synonyme: TCA
- CAS Nummer:76-03-9
- Formel:Cl₃CCOOH
- MDL-Nummer:MFCD00004177
- Molekulargewicht:163.39 g/mol
- Merck-Index:14,9627
- Siedepunkt:196 °C
- Schmelzender Platingehalt:54…58 °C
- Dichte:1,629
- Blitz Pt:113 °C (geschlossener Tiegel)
- Lagertemperatur:Raumtemperatur
- EINECS:200-927-2
- ADR:8,II
Dokumente
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1.000 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



