Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Caprylsäure ≥98%

Caprylsäure ≥98%

500 mL
Catalog # A11149.AP
Corrosive
Irritant
Caprylsäure ≥98%
Catalog # A11149.AP
CAS Number:  124-07-2
Caprylsäure ≥98%
Caprylsäure ≥98%

500 mL

CAS Number:  124-07-2
Price
SFr. 66.20
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 98+% CAS No.: 124-07-2 Molecular Formula: C8H16O2 Formula Weight: 144.21 Flash Point: 132°(269°F) Physical Form: Liquid Density: 0.910 Refractive Index: 1.4275 Appearance: Colourless Odour: Rancid pH: 4 Melting Point: 16-17° Boiling Point: 236-238° Beilstein Reference: 1747180 Merck Reference: 14,1765 UN No.: 3265 MDL No.: MFCD00004429

Synonyms:  Octanoic acid, n-Octanoic Acid, Octoic acid
  • CAS Number:124-07-2
  • Formula:C₈H₁₆O₂
  • MDL Number:MFCD00004429
  • Molecular Weight:144.21 g/mol
  • Merck Index:14,1765
  • Boiling Pt:236…238 °C
  • Melting Pt:16…17 °C
  • Density:0,910
  • Flash Pt:132 °C (269 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:204-677-5
  • UN:3265
  • ADR:8,III
Specifications

Specifications

  • VE:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.