Natriumpyruvat ≥99%
Glasflasche für Feststoffe, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Melting Point: 300° Beilstein Reference: 3568341
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Number:113-24-6
- Formula:CH₃COCOONa
- MDL Number:MFCD00002586
- Molecular Weight:110.04 g/mol
- Melting Pt:>300 °C
- Density:1,718 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:204-024-4
Documents
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



