Natriumpyruvat ≥99%
Glasflasche für Feststoffe, 250 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Melting Point: 300° Beilstein Reference: 3568341
Synonyme: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Nummer:113-24-6
- Formel:CH₃COCOONa
- MDL-Nummer:MFCD00002586
- Molekulargewicht:110.04 g/mol
- Schmelzender Platingehalt:>300 °C
- Dichte:1,718 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:204-024-4
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



