Natriumpyruvat ≥99%
Glasflasche für Feststoffe, 50 g
Purity: 99% CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Melting Point: 300° Beilstein Reference: 3568341
: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- :113-24-6
- :CH₃COCOONa
- :MFCD00002586
- :110.04 g/mol
- :>300 °C
- :1,718 g/cm³ (20 °C)
- :Raumtemperatur
- :204-024-4
- Packung:Glasflasche für Feststoffe
- VE:50 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



