4-Brombenzonitril ≥98%
Glasflasche für Feststoffe, 10 g
Purity: 98+% CAS No.: 623-00-7 Molecular Formula: C7H4BrN Formula Weight: 182.03 Flash Point: 130°(266°F) Melting Point: 111-114° Boiling Point: 235-237° Beilstein Reference: 2041518 UN No.: 3439 MDL No.: MFCD00001811
: p-Bromobenzonitrile
- :623-00-7
- :C₇H₄BrN
- :MFCD00001811
- :182.02 g/mol
- :235…237 °C
- :111…114 °C
- :1,562
- :130 °C (266 °F)
- :Raumtemperatur
- :210-764-9
- :3439
- :6.1,III
- Packung:Glasflasche für Feststoffe
- VE:10 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.