1,3-Propandiol ≥99%
Glasflasche für Feststoffe, 5.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 504-63-2 Molecular Formula: HO(CH2)3OH Formula Weight: 76.10 Flash Point: 140°(174°F) Refractive Index: 1.4400 Melting Point: -26° Boiling Point: 214° Solubility: Miscible with water, alcohol Beilstein Reference: 969155 Merck Reference: 14,9714 MDL No.: MFCD00002949
Synonyms: 1,3-Propylene glycol, 1,3-Dihydroxy propane
- CAS Number:504-63-2
- Formula:HO(CH₂)₃OH
- MDL Number:MFCD00002949
- Molecular Weight:76.1 g/mol
- Merck Index:14,9714
- Boiling Pt:214 °C
- Melting Pt:–26 °C
- Density:1,053
- Flash Pt:140 °C (174 °F)
- Storage Temperature:Raumtemperatur
- EINECS:207-997-3
Documents
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:5.000 g
- Cat. No.:A10829.0I
- Umweltzertifizierung:EPA Safer Chemical Solvent (Green Circle Rating)
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



