Phenoxyethanol ≥94%
Glass bottle for solids, 1.000 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- CAS Nummer:122-99-6
- Formel:C₆H₅OCH₂CH₂OH
- MDL-Nummer:MFCD00002857
- Molekulargewicht:138.17 g/mol
- Merck-Index:14,7257
- Siedepunkt:245 °C
- Schmelzender Platingehalt:11…14 °C
- Dichte:1,102
- Blitz Pt:121 °C (249 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:204-589-7
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1.000 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



