Methylhexanoat ≥99%
Glasflasche für Feststoffe, 250 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 106-70-7 Molecular Formula: CH3(CH2)4CO2CH3 Formula Weight: 130.19 Flash Point: 45°(113°F) Refractive Index: 1.4050 Melting Point: -71° Boiling Point: 150-151° Beilstein Reference: 1744683 UN No.: 3272 MDL No.: MFCD00009510
Synonyme: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
- CAS Nummer:106-70-7
- Formel:C₇H₁₄O₂
- MDL-Nummer:MFCD00009510
- Molekulargewicht:130.19 g/mol
- Siedepunkt:150…151 °C
- Schmelzender Platingehalt:–71 °C
- Dichte:0,885
- Blitz Pt:45 °C (113 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:203-425-1
- UN:3272
- ADR:3,III
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



