Nitrobenzol ≥99%
Glasflasche für Feststoffe, 10.000 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 98-95-3 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 87°(188°F) Refractive Index: 1.5520 Melting Point: 5-6° Boiling Point: 210-212° Beilstein Reference: 507540 Merck Reference: 14,6588 UN No.: 1662 MDL No.: MFCD00007043
- CAS Nummer:98-95-3
- Formel:C₆H₅NO₂
- MDL-Nummer:MFCD00007043
- Molekulargewicht:123.11 g/mol
- Merck-Index:14,6588
- Siedepunkt:210…212 °C
- Schmelzender Platingehalt:5…6 °C
- Dichte:1,196
- Blitz Pt:87 °C (188 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:202-716-0
- UN:1662
- ADR:6.1,II
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:10.000 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



