L(+)-Cystein ≥98%
Glasflasche für Feststoffe, 250 g
Purity: 98+% CAS No.: 52-90-4 Molecular Formula: C3H7NO2S Formula Weight: 121.16 Physical Form: Crystalline powder Sensitivity: Air Sensitive Appearance: White Odour: Odourless Melting Point: ca 220° dec. Beilstein Reference: 1721408 Merck Reference: 14,2781 Optical Rotation: +7.9° (c=5 in 5N HCl) MDL No.: MFCD00064306
: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
- :52-90-4
- :HSCH₂CH(NH₂)CO₂H
- :MFCD00064306
- :121.16 g/mol
- :14,2781
- :254 °C (1013 hPa)
- :∼ 220 °C (Zers.)
- :∼ 220 °C (Zersetzung)
- :1,523 g/cm³ (20 °C)
- :Raumtemperatur
- :200-158-2
Documents
- Packung:Glasflasche für Feststoffe
- VE:250 g