L(+)-Cystein ≥98%
Glasflasche für Feststoffe, 50 g
Purity: 98+% CAS No.: 52-90-4 Molecular Formula: C3H7NO2S Formula Weight: 121.16 Physical Form: Crystalline powder Sensitivity: Air Sensitive Appearance: White Odour: Odourless Melting Point: ca 220° dec. Beilstein Reference: 1721408 Merck Reference: 14,2781 Optical Rotation: +7.9° (c=5 in 5N HCl) MDL No.: MFCD00064306
: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
- :52-90-4
- :HSCH₂CH(NH₂)CO₂H
- :MFCD00064306
- :121.16 g/mol
- :14,2781
- :254 °C (1013 hPa)
- :∼ 220 °C (Zers.)
- :∼ 220 °C (Zersetzung)
- :1,523 g/cm³ (20 °C)
- :Raumtemperatur
- :200-158-2
- Packung:Glasflasche für Feststoffe
- VE:50 g
- Cat. No.:A10435.18
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.