Agarose
Glass bottle for solids, 500 g
Catalog # GRBU1036.0500
Marketsource
Supplier: GERBU BIOCHEMICALS
Jump to:
Product Details & Documents
- CAS Number:9012-36-6
- MDL Number:MFCD00081294
- Melting Pt:60…90 °C
- Storage Temperature:Raumtemperatur
- EINECS:232-731-8
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



