Natriumcarbonat ≥98%
Glasflasche für Feststoffe, 2.500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 497-19-8 Molecular Formula: Na2CO3 Formula Weight: 105.99 Refractive Index: 1.535 Odour: Odourless pH: 11,5 Beilstein Reference: 4154566 Merck Reference: 14,8596 MDL No.: MFCD00003494
Synonyme: Calcined soda, Carbonic acid disodium salt
- CAS Nummer:497-19-8
- Formel:Na₂CO₃
- MDL-Nummer:MFCD00003494
- Molekulargewicht:105.99 g/mol
- Merck-Index:14,8596
- Siedepunkt:1600 °C (1013 hPa)
- Schmelzender Platingehalt:854 °C
- Dichte:2,53
- Lagertemperatur:Raumtemperatur
- EINECS:207-838-8
- ERREICHEN:01-2119485498-19
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:2.500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



