Purity: 98% min CAS No.: 16940-66-2 Molecular Formula: BH4Na Formula Weight: 37.83 Physical Form: -4+16 Mesh Granules Density: 1.074 Appearance: White Odour: Weak Melting Point: 400° dec. Merck Reference: 14,8592 UN No.: 1426 MDL No.: MFCD00003518
: Sodium tetrahydridoborate, Sodium tetrahydroborate
- :16940-66-2
- :NaBH₄
- :MFCD00003518
- :37.83 g/mol
- :14,8592
- :400 °C (Zers.)
- :400 °C (Zersetzung)
- :1,074
- :69 °C
- :Raumtemperatur
- :241-004-4
- :1426
- :4.3,I
- Packung:Glass bottle for solids
- VE:10 g