n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
5 L
Anbieter: Avantor
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Katalognr. 8044.5000
Anbieter: Avantor
CAS Nummer: 110-54-3
Direkt zu
Product Details & Documents
Synonyme: Hexyl hydride, Hexanen, Hexane
- CAS Nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL-Nummer:MFCD00009520
- Molekulargewicht:86.18 g/mol
- Merck-Index:13,04712
- Siedepunkt:69 °C (1013 hPa)
- Schmelzender Platingehalt:–94,3 °C
- Dichte:0,659 g/cm³ (20 °C)
- Blitz Pt:–22 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
Ergebnisse des Spezifikationstests
- Assay (by GC):min. 95 %
- Aromatic Compounds (as C6H6):max. 0,02 %
- Color (APHA):max. 10
- Density (g/ml) at 25°C:max. 0,657
- Residue after Evaporation:max. 0,001 %
- Sulfur Compounds (as S):max. 0,005 %
- Thiophene:passes test
- Water Soluble Titrable Acid (meq/g):max. 0,0003
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Technische Daten
- VE:5 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



