Sulfolan ≥99%
Glass bottle for solids, 250 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 126-33-0 Molecular Formula: C4H8O2S Formula Weight: 120.17 Flash Point: 166°(330°F) Refractive Index: 1.4825 Odour: Weak, characteristic Melting Point: 25-28° Boiling Point: 285° Beilstein Reference: 107765 Merck Reference: 14,8955 MDL No.: MFCD00005484
Synonyme: Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene 1,1-dioxide
- CAS Nummer:126-33-0
- Formel:C₄H₈O₂S
- MDL-Nummer:MFCD00005484
- Molekulargewicht:120.17 g/mol
- Merck-Index:14,8955
- Siedepunkt:285 °C
- Schmelzender Platingehalt:25…28 °C
- Dichte:1,261
- Blitz Pt:166 °C (330 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:204-783-1
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:250 g



