AuftragsverfolgungKontakt
Kryoboxen
Kryoboxen
100 (10×10), up to 10 mm, 134×134×57 mm
Catalog # DELTM-606
Supplier:   DELTALAB
Kryoboxen
Catalog # DELTM-606
Supplier:  DELTALAB
Kryoboxen
100 (10×10), up to 10 mm, 134×134×57 mm
Price
SFr. 239.00
Log in for availability
1 * 10 ST
Product Details & Documents
Cryoboxes, cardboard, white, with pre-assembled compartments are designed for storage and preservation of frozen samples.
  • Covered by special white paper hand-stuck and hand-assembled for a better quality
  • Resist temperatures up to –100 °C
  • Alphanumeric identifications printed on each box
Suitable for the racks of freezer chests. Ideal for microtubes and cryovials.
Specifications
  • Fächer:
    100 (10×10)
  • Für Röhrchen Ø:
    up to 10 mm
  • Außen B×T×H:
    134×134×57 mm
  • Seq:
    03