Tube racks, universal
Pink, 32/32/12/4, 0,5/1,5-2,0/15/50 ml, PP, 172×50×90 mm
Katalognr. DELTM-522
Anbieter: DELTALAB
Direkt zu
Product Details & Documents
PP, linkable to other racks.
- Versatile rack with different capacities on each of the four sides
- Alpha-numeric matrix on the four sides for better sample location
- Suitable for freezers and autoclaves
Dokumente
Zertifikate
Suche nach ChargennummerTechnische Daten
- Für Röhrchen:0,5/1,5-2,0/15/50 ml
- Anz. Bohrungen:32/32/12/4
- Farbe:Pink
- Material:PP
- Seq:02
- B×T×H:172×50×90 mm