Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
PIG® SUPER absorbent socks | Avantor

PIG® SUPER absorbent socks

Absorbent socks, PP skin/flame-retardant cellulose filler, 91,2 L, 24 socks/box, 83×1070 mm
Katalognr. 121-0537
Anbieter:   NEW PIG
PIG® SUPER absorbent socks
Katalognr. 121-0537
Anbieter:  NEW PIG
PIG® SUPER absorbent socks
PIG® SUPER absorbent socks

Absorbent socks, PP skin/flame-retardant cellulose filler, 91,2 L, 24 socks/box, 83×1070 mm

Preis
SFr. 147.00
Einloggen für Verfügbarkeit
1 * 24 ST
Product Details & Documents

PIG® SUPER Absorbent Sock is great for high-volume machine leaks. Soaks up nearly twice as much liquid as other maintenance socks.

  • Extra-thirsty, formable socks hug corners and walls, surround higher-volume machine leaks and contain spills to keep workers safe
  • Cellulose filler is highly absorbent for big leaks and spills; super absorbency means that you replace them less often
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water; not recommended for acids, caustics or other corrosive liquids
  • 85% recycled cellulose filler is ideal when green products are desired

Wringable for reclaiming up to 65% of absorbed liquid. Can be incinerated after use to reduce waste or for fuels blending.

Technische Daten
  • Bezeichnung:
    Absorbent socks, PP skin/flame-retardant cellulose filler
  • Seq:
    02
  • Absorptionsvermögen:
    91,2 L
  • Verpackt:
    24 socks/box
  • Ø×L:
    83×1070 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.