AuftragsverfolgungKontakt
Stoppers for Conical Ground Joints
Stoppers for Conical Ground Joints
29/32, Flat
Katalognr. LENZ01400329
Anbieter:   Lenz Laborglas
Stoppers for Conical Ground Joints
Katalognr. LENZ01400329
Anbieter:  Lenz Laborglas
Stoppers for Conical Ground Joints
Stoppers for Conical Ground Joints
29/32, Flat
Preis
SFr. 108.00
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certifications: DIN 12252
Ordering information: Available with flat or pointed bottom.
Documents
Technische Daten
  • NS:
    29/32
  • Typ:
    Flat
  • Seq:
    16