Standard ground joints
12/21 NS, - NS, 11 mm
Catalog # QUTW1043
Supplier: QUARTZTECHNISCHE WERKSTATTEN
Jump to:
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving high resistance to temperature changes.
Specifications
- Kern:12/21 NS
- Hülse:- NS
- Rohr A-Ø:11 mm
- Seq:05