Solid phase extraction cartridges, Bond Elut™ C18 EWP
LRC-Kartuschen, C18 EWP, 500 Å, 40 µm, 10 ml, 500 mg
Catalog # VARI12113071
Supplier: VARIAN
Jump to:
Product Details & Documents
Bond Elut™ EWP is based upon standard particle size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15000 MW), which are typically excluded from standard porosity silica phases.
- No exclusion of large molecules
- Good for desalting proteins
- Successful separation of proteins, peptides or nucleotides
Specifications
- Partikelgröße:40 µm
- Porengröße:500 Å
- Kapazität:10 ml
- Bezeichnung:LRC-Kartuschen
- Bettgewicht:500 mg
- Phase:C18 EWP
- Cat. No.:VARI12113071
- Endcapped:Ja
- Kohlenstoffbelastung:6,0%
- Surface area:80 m²/g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



