Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Solid phase extraction cartridges, Bond Elut™ AccuCAT | Avantor

Solid phase extraction cartridges, Bond Elut™ AccuCAT

Straight barrel cartridge, AccuCAT, 40 µm, 6 ml, 400 mg
Katalognr. VARI12282006
Anbieter:   VARIAN
Solid phase extraction cartridges, Bond Elut™ AccuCAT
Katalognr. VARI12282006
Anbieter:  VARIAN
Solid phase extraction cartridges, Bond Elut™ AccuCAT
Solid phase extraction cartridges, Bond Elut™ AccuCAT

Straight barrel cartridge, AccuCAT, 40 µm, 6 ml, 400 mg

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 30 ST
Product Details & Documents

Bond Elut AccuCAT cartridges are mixed bed SPE cartridges consisting of a strong cation exchange (SCX) and a strong anion exchange (SAX) sorbent packed into one bed. AccuCAT is effective for the extraction of acidic, basic and neutral analytes from urine and other biological samples. AccuCAT is particularly effective for catecholamine extraction from bio fluids.

  • Ultra clean, mixed sorbent bed delivers for reproducible extractions
  • SCX and SAX functionalities offer broad analyte extraction potential
  • Compatible with many biological fluids for easy method transfer
Technische Daten
  • Partikelgröße:
    40 µm
  • Kapazität:
    6 ml
  • Bezeichnung:
    Straight barrel cartridge
  • Seq:
    06
  • Bettgewicht:
    400 mg
  • Phase:
    AccuCAT
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.