Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Pergolide Related Compounds, Reference Standard | Avantor

Pergolide Related Compounds, Reference Standard

Pergolide Mesylate, USP reference standard, Vial, 200 mg
Katalognr. 1510845.
Anbieter:   USP
Pergolide Related Compounds, Reference Standard
Katalognr. 1510845.
Anbieter:  USP
CAS Nummer:  
Pergolide Related Compounds, Reference Standard

Pergolide Mesylate, USP reference standard, Vial, 200 mg

CAS Nummer:  
Preis
SFr. 499.00
Einloggen für Verfügbarkeit
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Zertifizierung: US Pharmacopea Reference Standard

Technische Daten

Technische Daten

  • Packung:
    Vial
  • VE:
    200 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Pergolide Mesylate
  • UN No.:
    1544
  • Cat. No.:
    1510845.
  • CAS Nr.:
    66104-23-2
  • Bezeichnung:
    Pergolide Mesylate

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.