Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Acetophenon ≥99%

Acetophenon ≥99%

Glasflasche, 500 mL
Catalog # A12727.AP
Irritant
Acetophenon ≥99%
Catalog # A12727.AP
CAS Number:  98-86-2
Acetophenon ≥99%
Acetophenon ≥99%

Glasflasche, 500 mL

CAS Number:  98-86-2
Price
SFr. 58.20
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 98-86-2 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 76°(169°F) Refractive Index: 1.5325 Melting Point: 19-20° Boiling Point: 201-203° Merck Reference: 14,73 MDL No.: MFCD00008724

Synonyms:  Methyl phenyl ketone, Phenyl methyl ketone
  • CAS Number:98-86-2
  • Formula:C₈H₈O
  • MDL Number:MFCD00008724
  • Molecular Weight:120.15 g/mol
  • Merck Index:14,73
  • Boiling Pt:201…203 °C
  • Melting Pt:19…20 °C
  • Density:1,030
  • Flash Pt:76 °C (169 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:202-708-7
  • UN:3334
  • ADR:9,
Specifications

Specifications

  • Packung:
    Glasflasche
  • VE:
    500 mL

Specifications