Capillary column ferrules for gas chromatography
High temperature PTV inlet, SS/graphite ferrule ID 0,4 mm, column ID 0,32 mm
Katalognr. HEWL5188-5315
Anbieter: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalognr. HEWL5188-5315
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datasheet
Technische Daten
- Bezeichnung:High temperature PTV inlet, SS/graphite ferrule ID 0,4 mm, column ID 0,32 mm
- Seq:21