Electron capture detectors (ECD)
Micro ECD make-up gas adapter
Catalog # HEWLG2397-80520
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
- Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Specifications
- Bezeichnung:Micro ECD make-up gas adapter
- Seq:07