CrossLab Bottle Head Assemblies for HPLC
Frit adapter, PTFE
Catalog # HEWLG2258-23201
Supplier: AGILENT HEWLETT PACKARD
CrossLab Bottle Head Assemblies for HPLC
Catalog # HEWLG2258-23201
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
All the parts required to connect solvent bottles to your HPLC instrument.
- Instrument specific ready-to-use bottle head assemblies available
- Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.
Specifications
- Bezeichnung:Frit adapter, PTFE
- Seq:12
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.