α-Linolensäure
Glass bottle for solids, 100 mg
Katalognr. CAYM90210-100
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
Synonyme: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid
- CAS Nummer:463-40-1
- Formel:C₁₈H₃₀O₂
- MDL-Nummer:MFCD00065720
- Molekulargewicht:278.44 g/mol
- Merck-Index:12,05530
- Siedepunkt:443 °C (1013 hPa)
- Schmelzender Platingehalt:–9,5 °C
- Dichte:0,9141 g/cm³ (18 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:207-334-8
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



