Test tubes, disposable, without rim
160 mm, 16 mm, 0,5 mm
Katalognr. HECH42769051
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
Glas, clear.
- Excellent chemical resistance
- Good temperature resistance
Technische Daten
- Ext.Ø:16 mm
- Wandstärke:0,5 mm
- Länge:160 mm