Tanks for water baths
Plexiglas® tank with inclined bottom, 430×130×150 mm
Katalognr. HECH43187003
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
Tubs made of Plexiglas®, available with or without sloping base. Sloping base facilitates circulation of the liquid.
For use with water bath thermostats.
Technische Daten
- Bezeichnung:Plexiglas® tank with inclined bottom, 430×130×150 mm
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.