Clips for conical joints
Orange, 34
Catalog # ROTH1169.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
These clips for conical joints are made of POM.
- Resistant to most chemicals
- Temperature stable up to 90 °C
Delivery information: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Specifications
- Für NS:34
- Farbe:Orange
- Seq:07