AuftragsverfolgungKontakt
Clips for conical joints | Avantor
Clips for conical joints
Blau, 19
Katalognr. ROTH1166.1
Marketsource
Anbieter:   Roth Carl
Clips for conical joints
Katalognr. ROTH1166.1
Anbieter:  Roth Carl
Marketsource
Clips for conical joints
Blau, 19
Preis
60.50 SFr.
Einloggen für Verfügbarkeit
1 * 12 ST
Product Details & Documents
These clips for conical joints are made of POM.
  • Resistant to most chemicals
  • Temperature stable up to 90 °C
 : Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Technische Daten
  • Für NS:
    19
  • Farbe:
    Blau
  • Seq:
    04