Clips for conical joints
Gelb, 14
Katalognr. ROTH1165.1
Marketsource
Anbieter: Roth Carl
Direkt zu
Product Details & Documents
These clips for conical joints are made of POM.
- Resistant to most chemicals
- Temperature stable up to 90 °C
Lieferinformationen: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
Technische Daten
- Für NS:14
- Farbe:Gelb
- Seq:03