Clips for conical joints
Lila, 12
ROTH1164.1
Marketsource
These clips for conical joints are made of POM.
- Resistant to most chemicals
- Temperature stable up to 90 °C
: Assorted colour pack contains one of each colour (blue, green, violet, yellow, brown and orange).
- Für NS:12
- Farbe:Lila
- Seq:02