Bicin
Glasflasche für Feststoffe, 50 g
Catalog # ROTH9162.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
- CAS Number:150-25-4
- Formula:C₆H₁₃NO₄
- MDL Number:MFCD00004295
- Molecular Weight:163.17 g/mol
- Melting Pt:185…192 °C
- Storage Temperature:Raumtemperatur
- EINECS:205-755-1
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:50 g