Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Natriumpyruvat

Natriumpyruvat

Glasflasche für Feststoffe, 250 g
Catalog # ROTH8793.2
Marketsource
Supplier:   Roth Carl
Natriumpyruvat
Catalog # ROTH8793.2
Supplier:  Roth Carl
Marketsource
CAS Number:  113-24-6
Natriumpyruvat

Glasflasche für Feststoffe, 250 g

CAS Number:  113-24-6
Price
123.00 SFr.
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  • CAS Number:113-24-6
  • Formula:CH₃COCOONa
  • MDL Number:MFCD00002586
  • Molecular Weight:110.04 g/mol
  • Melting Pt:220…230 °C
  • Density:1,718 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:204-024-4
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.