Trichloressigsäure
Glasflasche für Feststoffe, 500 g
ROTH8789.2
Marketsource
: TCA
- :76-03-9
- :Cl₃CCOOH
- :MFCD00004177
- :163.39 g/mol
- :13,09700
- :196 °C (1013 hPa)
- :54…56 °C
- :1,63 g/cm³ (20 °C)
- :113 °C (geschlossener Tiegel)
- :Raumtemperatur
- :200-927-2
- Packung:Glasflasche für Feststoffe
- VE:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.