Natriumcarbonat
10 kg
Katalognr. ROTH8563.2
Marketsource
Anbieter: Roth Carl
Direkt zu
Product Details & Documents
Synonyme: Calcined soda, Carbonic acid disodium salt
- CAS Nummer:497-19-8
- Formel:Na₂CO₃
- MDL-Nummer:MFCD00003494
- Molekulargewicht:105.99 g/mol
- Merck-Index:14,08596
- Siedepunkt:1600 °C (1013 hPa)
- Schmelzender Platingehalt:854 °C
- Dichte:2,53 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:207-838-8
- ERREICHEN:01-2119485498-19
Technische Daten
Technische Daten
- VE:10 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



