Natriumcarbonat
Glass bottle for solids, 500 g
ROTHA135.1
Marketsource
: Calcined soda, Carbonic acid disodium salt
- :497-19-8
- :Na₂CO₃
- :MFCD00003494
- :105.99 g/mol
- :14,08596
- :1600 °C (1013 hPa)
- :854 °C
- :2,53 g/cm³ (20 °C)
- :Raumtemperatur
- :207-838-8
- :01-2119485498-19
- Packung:Glass bottle for solids
- VE:500 g