Essigsäure 60%
Glasflasche, 1 L
Katalognr. ROTH7329.1
Marketsource
Anbieter: Roth Carl
Direkt zu
Product Details & Documents
Synonyme: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Nummer:64-19-7
- Formel:H₃CCOOH
- MDL-Nummer:MFCD00036152
- Molekulargewicht:60.05 g/mol
- Lagertemperatur:Raumtemperatur
- EINECS:200-580-7
- UN:2790
- ADR:8,II
Technische Daten
Technische Daten
- Packung:Glasflasche
- VE:1 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



