Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)

MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)

Glass bottle for solids, 100 g
Catalog # ROTH7117.2
Irritant
Marketsource
Supplier:   Roth Carl
MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)
Catalog # ROTH7117.2
Supplier:  Roth Carl
Marketsource
CAS Number:  68399-77-9
MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)

Glass bottle for solids, 100 g

CAS Number:  68399-77-9
Price
202.80 SFr.
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Hydroxy-3-morpholinopropane-1-sulphonic acid
  • CAS Number:68399-77-9
  • Formula:C₇H₁₅NO₅S
  • MDL Number:MFCD00009629
  • Molecular Weight:225.27 g/mol
  • Storage Temperature:Raumtemperatur
  • EINECS:269-989-6
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    100 g

Specifications